Learn More
Medchemexpress LLC HY-120697 5mg Medchemexpress, MSAB CAS:173436-66-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1206975MG
Encompass_Preferred
Medchemexpress, HY-120697 5mg MSAB CAS:173436-66-3 MSAB is a potent and selective inhibitor of Wnt/β-catenin signaling. MSAB binds to β-catenin promoting its degradation, and specifically downregulates Wnt/β-catenin target genes. MSAB exhibits potent anti-tumor effects selectively on Wnt-dependent cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0950