Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MSC 2032964A Inhibitor
SDP

Catalog No. NB564150MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB564150MG 50 mg
NB564110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB564150MG Supplier Novus Biologicals™ Supplier No. NB564150MG

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Low molecular weight, organic compound that may help regulate biological process.

Biological Activity: Potent and selective ASK1 inhibitor (IC50 = 93 nM). Selective for ASK1 over a panel of 210 kinases. Blocks LPS-induced ASK1 and p38 phosphorylation in cultured mouse astrocytes. Suppresses neuroinflammation in a mouse EAE model. Orally bioavailable and brain penetrant.

Storage: Store at 4°C.

Note: Inhibitors are guaranteed for 1 year from date of receipt.

Specifications

Product Type Inhibitor
Target Proteins ASK1
CAS 1124381-43-6
Molecular Formula C16H13F3N6O
Formula Weight 362.41g/mol
Content And Storage Store at 20°C
For Use With (Application) Potent and selective ASK1 inhibitor
Quantity 50 mg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.