Learn More
Promocell GmbH PromoCell Mesenchymal Stem Cell Adipogenic Different. Medium 2 with C-39816 attached to flask 100 ml, ready-to-use
Catalog No. NC1168186
Encompass
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).PromoCell offers five MSC Differentiation Media to efficiently induce differentiation of MSC into adipogenic, chondrogenic (w/ and w/o inducers), osteogenic or neurogenic lineages, respectively.PromoCell Mesenchymal Stem Cell Adipogenic Differentiation Media was developed for the directed differentiation of mesenchymal stem cells (MSC from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into adipogenic lineages.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.
Catalog No. NC1168186 Supplier Promocell GmbH Supplier No. C28016
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).PromoCell offers five MSC Differentiation Media to efficiently induce differentiation of MSC into adipogenic, chondrogenic (w/ and w/o inducers), osteogenic or neurogenic lineages, respectively.PromoCell Mesenchymal Stem Cell Adipogenic Differentiation Media was developed for the directed differentiation of mesenchymal stem cells (MSC from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into adipogenic lineages.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.