Learn More
MSC MSC Aircraft Cable, 1/4 in, 6 x 37 IWRC, Galvanized
Catalog No. 5000531653
Encompass
This aircraft cable from MSC Industrial Supply Co. is part of the Value Collection, offering quality at a great value. It features a 1/4 in diameter and 6 x 37 IWRC strand type, with a galvanized finish for enhanced durability. This cut-to-length item is designed for various applications requiring high strength, such as general utility and rigging.
  • Aircraft cable construction for general utility and rigging.
  • 1/4 in diameter.
  • 6 x 37 IWRC strand type.
  • Galvanized finish for corrosion resistance.
  • 5300 lb breaking strength.
  • Uncoated head.
  • Offers quality and value.
  • Can be custom cut to length.
Catalog No. 50-005-31653 Supplier MSC Supplier No. 89489066
This aircraft cable from MSC Industrial Supply Co. is part of the Value Collection, offering quality at a great value. It features a 1/4 in diameter and 6 x 37 IWRC strand type, with a galvanized finish for enhanced durability. This cut-to-length item is designed for various applications requiring high strength, such as general utility and rigging.
  • Aircraft cable construction for general utility and rigging.
  • 1/4 in diameter.
  • 6 x 37 IWRC strand type.
  • Galvanized finish for corrosion resistance.
  • 5300 lb breaking strength.
  • Uncoated head.
  • Offers quality and value.
  • Can be custom cut to length.