Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC MSC Amara Mechanic and Lifting Gloves, Black, Medium, 1 Pair

Catalog No. 5000684655
Encompass

These MSC gloves, size medium, are designed for Mechanic's & Lifting applications, providing both impact and anti-vibration protection. Crafted from durable Amara material, they feature a convenient Hook & Loop cuff for a secure fit. The full-fingered design ensures comprehensive coverage, and they come in a sleek black color. Ideal for tasks requiring robust hand protection and comfort.

  • Application: Mechanic's & Lifting
  • Cuff Style: Hook & Loop
  • Primary Material: Amara
  • Glove Size: Medium (Numeric Size: 8)
  • Color: Black
  • Protection Type: Impact; Anti-Vibration
  • Hand: Pair
  • Finger Coverage: Full Fingered

Catalog No. 50-006-84655 Supplier MSC Supplier No. 04978441
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These MSC gloves, size medium, are designed for Mechanic's & Lifting applications, providing both impact and anti-vibration protection. Crafted from durable Amara material, they feature a convenient Hook & Loop cuff for a secure fit. The full-fingered design ensures comprehensive coverage, and they come in a sleek black color. Ideal for tasks requiring robust hand protection and comfort.

  • Application: Mechanic's & Lifting
  • Cuff Style: Hook & Loop
  • Primary Material: Amara
  • Glove Size: Medium (Numeric Size: 8)
  • Color: Black
  • Protection Type: Impact; Anti-Vibration
  • Hand: Pair
  • Finger Coverage: Full Fingered

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.