Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC MSC Anti-Vibration Gloves, Mechanic and Lifting, Large, 1 Pair

Catalog No. 5000687157
Encompass

These work and general purpose gloves are designed for mechanic's and lifting applications. They are black and sold as a pair, offering reliable hand protection.

  • Application: mechanic's and lifting
  • Cuff style: hook and loop
  • Primary material: Amara
  • Color: black
  • High visibility: no
  • Static dissipative: no
  • Hand: pair
  • Finger coverage: full fingered
  • Protection type: impact and anti-vibration

Catalog No. 50-006-87157 Supplier MSC Supplier No. 04978466
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These work and general purpose gloves are designed for mechanic's and lifting applications. They are black and sold as a pair, offering reliable hand protection.

  • Application: mechanic's and lifting
  • Cuff style: hook and loop
  • Primary material: Amara
  • Color: black
  • High visibility: no
  • Static dissipative: no
  • Hand: pair
  • Finger coverage: full fingered
  • Protection type: impact and anti-vibration

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.