Learn More
Aviva Systems Biology Corp Msc Antibody - C-terminal region (ARP37092_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37092_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Msc. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of mouse Msc. Alternate names: MyoR, bHLHa22ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-584