Learn More
MSC MSC AR Gloves, Cut A4, 3X-Large, 1 ea
Catalog No. 5000677535
Encompass
AR Top-Grain Cowhide Leather Driver's Gloves provide arc flash, cut, and puncture resistance, designed for comfort and durability in demanding work environments.
  • Split-cowhide leather back provides comfort, durability, abrasion resistance, and breathability.
  • Top-grain cowhide leather palm is smooth and provides durability and dexterity.
  • Para-aramid blend lining provides added cut and heat resistance.
  • Keystone thumb is ergonomic to reduce hand fatigue.
  • Sewn with Kevlar® for maximum seam strength.
  • Slip-on styling to provide good fit and comfort.
  • Arc Rating 34 cal/cm.
  • Ansi Cut Level A4.
  • Ansi Puncture Level 3.
Catalog No. 50-006-77535 Supplier MSC Supplier No. 06004907
AR Top-Grain Cowhide Leather Driver's Gloves provide arc flash, cut, and puncture resistance, designed for comfort and durability in demanding work environments.
  • Split-cowhide leather back provides comfort, durability, abrasion resistance, and breathability.
  • Top-grain cowhide leather palm is smooth and provides durability and dexterity.
  • Para-aramid blend lining provides added cut and heat resistance.
  • Keystone thumb is ergonomic to reduce hand fatigue.
  • Sewn with Kevlar® for maximum seam strength.
  • Slip-on styling to provide good fit and comfort.
  • Arc Rating 34 cal/cm.
  • Ansi Cut Level A4.
  • Ansi Puncture Level 3.