Learn More
Promocell GmbH PromoCell Mesenchymal Stem Cell Chondrogenic Different. Medium 100 ml, ready-to-use, with C-39812 attached to flask
Catalog No. 50306279
Encompass
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).PromoCell offers five MSC Differentiation Media to efficiently induce differentiation of MSC into adipogenic, chondrogenic (w/ and w/o inducers), osteogenic or neurogenic lineages, respectively.PromoCell Mesenchymal Stem Cell Chondrogenic Differentiation Media is a serum-free medium developed for the directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into chondrogenic lineages.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.
Catalog No. 50-306-279 Supplier Promocell GmbH Supplier No. C28012
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).PromoCell offers five MSC Differentiation Media to efficiently induce differentiation of MSC into adipogenic, chondrogenic (w/ and w/o inducers), osteogenic or neurogenic lineages, respectively.PromoCell Mesenchymal Stem Cell Chondrogenic Differentiation Media is a serum-free medium developed for the directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into chondrogenic lineages.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.