Learn More
MSC MSC Duct Elbow, Galvanized, 90 Degree, 4 in, 1 Each
Catalog No. 5000504187
Encompass
This galvanized duct fitting is a 90 degree elbow designed for industrial ventilation and dust collection systems. It features a 4 inch inside diameter and a durable 24 gage construction, ensuring robust performance and longevity in demanding environments.
  • Ninety degree elbow design
  • Four inch inside diameter
  • Seven point thirty-two inch overall length
  • Galvanized material for corrosion resistance
  • Made in USA
Catalog No. 50-005-04187 Supplier MSC Supplier No. 03233087
This galvanized duct fitting is a 90 degree elbow designed for industrial ventilation and dust collection systems. It features a 4 inch inside diameter and a durable 24 gage construction, ensuring robust performance and longevity in demanding environments.
  • Ninety degree elbow design
  • Four inch inside diameter
  • Seven point thirty-two inch overall length
  • Galvanized material for corrosion resistance
  • Made in USA