Learn More
Promocell GmbH PromoCell MSC Growth Medium XF 500 ml, ready-to-use, with C-39819 attached to flask
Catalog No. 50306283
Encompass
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).The PromoCell MSC Growth Media are designed to robustly support the standardized expansion of multipotent human Mesenchymal Stem Cells. Rigorous quality control tests ensure reliable and consistent product quality.The Mesenchymal Stem Cell Growth Medium XF (xeno-free) allows for the robust proliferation of multipotent MSC in a serum-free and xeno-free culture environment. Fibronectin-coated plates are necessary in conjunction with this medium.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.
Catalog No. 50-306-283 Supplier Promocell GmbH Supplier No. C28019
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).The PromoCell MSC Growth Media are designed to robustly support the standardized expansion of multipotent human Mesenchymal Stem Cells. Rigorous quality control tests ensure reliable and consistent product quality.The Mesenchymal Stem Cell Growth Medium XF (xeno-free) allows for the robust proliferation of multipotent MSC in a serum-free and xeno-free culture environment. Fibronectin-coated plates are necessary in conjunction with this medium.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.