Learn More
MSC MSC Industrial Supply Co. Clevis Hook, 9/32 in Chain Diameter, Grade 100, 4,300 Lb Capacity
Catalog No. 5000528697
Encompass
This clevis hook is designed for heavy-duty applications, featuring a 9/32 inch chain diameter and a Grade 100 rating for robust performance. Constructed from durable alloy steel, it offers a substantial 4,300 Lb capacity, making it suitable for various lifting and rigging tasks.
  • Chain diameter: 9/32 in
  • Chain grade: 100
  • Material: Alloy steel
  • Capacity: 4,300 Lb
  • Internal diameter (ID): 11/16 in
  • Pin diameter: 13/32 in
  • Hook throat: 3/8 in
  • Overall length (OAL): 2-1/8 in
Catalog No. 50-005-28697 Supplier MSC Supplier No. 81007304
This clevis hook is designed for heavy-duty applications, featuring a 9/32 inch chain diameter and a Grade 100 rating for robust performance. Constructed from durable alloy steel, it offers a substantial 4,300 Lb capacity, making it suitable for various lifting and rigging tasks.
  • Chain diameter: 9/32 in
  • Chain grade: 100
  • Material: Alloy steel
  • Capacity: 4,300 Lb
  • Internal diameter (ID): 11/16 in
  • Pin diameter: 13/32 in
  • Hook throat: 3/8 in
  • Overall length (OAL): 2-1/8 in