Learn More
MSC MSC Industrial Supply Co. Detachable Sleeve, Brass, Nickel Plated, 9/32 Inch Long
Catalog No. 5000513242
Encompass
This detachable sleeve is designed for use with ball chains, offering reliable performance in various applications. It is constructed from durable brass with a nickel-plated finish, providing enhanced resistance and longevity. Measuring 9/32 inch in length, this sleeve is compatible with trade size numbers 6 and 10 ball chains.
  • Constructed from brass for durability.
  • Nickel plated finish for corrosion resistance.
  • Measures 9/32 inch in length.
  • Compatible with trade size numbers 6 and 10 ball chains.
  • Made in USA.
Catalog No. 50-005-13242 Supplier MSC Supplier No. 01727304
This detachable sleeve is designed for use with ball chains, offering reliable performance in various applications. It is constructed from durable brass with a nickel-plated finish, providing enhanced resistance and longevity. Measuring 9/32 inch in length, this sleeve is compatible with trade size numbers 6 and 10 ball chains.
  • Constructed from brass for durability.
  • Nickel plated finish for corrosion resistance.
  • Measures 9/32 inch in length.
  • Compatible with trade size numbers 6 and 10 ball chains.
  • Made in USA.