Learn More
MSC MSC Industrial Supply Co. End Coupling, Nickel Plated Brass, 13/32 Inch Long, For #3 and #6 Ball Chain
Catalog No. 5000529774
Encompass
This end coupling is constructed from nickel plated brass and measures 13/32 inch long. It is designed for securing #3 and #6 size ball chains, making it suitable for various industrial and commercial applications.
  • Constructed from nickel plated brass.
  • Measures 13/32 inch long.
  • Compatible with #3 and #6 size ball chains.
  • Available in packs of 100.
  • Manufactured in the USA.
  • Returnable within 30 days of shipment in original packaging if expectations are not met (excluding hazardous materials and cut-to-length products).
Catalog No. 50-005-29774 Supplier MSC Supplier No. 01727213
This end coupling is constructed from nickel plated brass and measures 13/32 inch long. It is designed for securing #3 and #6 size ball chains, making it suitable for various industrial and commercial applications.
  • Constructed from nickel plated brass.
  • Measures 13/32 inch long.
  • Compatible with #3 and #6 size ball chains.
  • Available in packs of 100.
  • Manufactured in the USA.
  • Returnable within 30 days of shipment in original packaging if expectations are not met (excluding hazardous materials and cut-to-length products).