Learn More
MSC MSC Industrial Supply Co. Guidance Pulley, Single Side, Side Mount, Malleable Iron, Galvanized
Catalog No. 5000531038
Encompass
This guidance pulley features a 1-1/2 inch outside diameter sheave, designed for use with 3/8 inch diameter fibrous ropes. Constructed from malleable iron with a galvanized finish, it is suitable for various applications requiring smooth rope guidance.
  • Sheave with 1-1/2 inch outside diameter
  • Suitable for fibrous ropes up to 3/8 inch in diameter
  • Mounting hole diameter ranges from 0.183 inch to 0.194 inch
  • Made from durable malleable iron
  • Galvanized finish for enhanced protection
Catalog No. 50-005-31038 Supplier MSC Supplier No. 67328625
This guidance pulley features a 1-1/2 inch outside diameter sheave, designed for use with 3/8 inch diameter fibrous ropes. Constructed from malleable iron with a galvanized finish, it is suitable for various applications requiring smooth rope guidance.
  • Sheave with 1-1/2 inch outside diameter
  • Suitable for fibrous ropes up to 3/8 inch in diameter
  • Mounting hole diameter ranges from 0.183 inch to 0.194 inch
  • Made from durable malleable iron
  • Galvanized finish for enhanced protection