Learn More
MSC MSC Industrial Supply Co. Plug & Pin Gage, Class ZZ Minus, 0.4410 in Diameter, Steel
Catalog No. 5000528808
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.4410 inch diameter and is constructed from durable steel. It is designed for precise measurement applications, offering a tight tolerance of 0.000200 "
  • In stock
  • Delivered as soon as next business day
  • Hassle free guarantee: we will exchange or refund any product that does not meet your expectations
Catalog No. 50-005-28808 Supplier MSC Supplier No. 78376373
This Class ZZ Minus Plug & Pin Gage features a 0.4410 inch diameter and is constructed from durable steel. It is designed for precise measurement applications, offering a tight tolerance of 0.000200 "
  • In stock
  • Delivered as soon as next business day
  • Hassle free guarantee: we will exchange or refund any product that does not meet your expectations