Learn More
MSC MSC Industrial Supply Co. Plumber's Safety Chain, Brass, 0.023 in, 40 lb
Catalog No. 5000531050
Encompass
This brass plumber's safety chain features weldless construction, a 0.023 inch diameter, and a 40 lb work load limit, designed for reliable performance in plumbing applications.
  • Brass material for durability and corrosion resistance.
  • Weldless construction ensures strength and safety.
  • Standard finish for general applications.
  • 0.023 inch diameter and 100 ft overall length.
  • 40 lb work load limit for secure fastening.
  • Features 24.7 links per foot.
  • Inside length of 0.55 "
Catalog No. 50-005-31050 Supplier MSC Supplier No. 67776229
This brass plumber's safety chain features weldless construction, a 0.023 inch diameter, and a 40 lb work load limit, designed for reliable performance in plumbing applications.
  • Brass material for durability and corrosion resistance.
  • Weldless construction ensures strength and safety.
  • Standard finish for general applications.
  • 0.023 inch diameter and 100 ft overall length.
  • 40 lb work load limit for secure fastening.
  • Features 24.7 links per foot.
  • Inside length of 0.55 "