Learn More
MSC MSC Industrial Supply Co. Rigid Eye Single Sheave Pulley, 1.5 in Outside Dia, Malleable Iron, Galvanized
Catalog No. 5000530609
Encompass
This Rigid Eye Single Sheave Pulley is designed for applications utilizing fibrous ropes. It features a 1.5 in outside diameter and is constructed from durable malleable iron with a galvanized finish, ensuring quality performance for various business needs.
  • Designed for use with fibrous ropes
  • Features a 1.5 in outside diameter
  • Constructed from malleable iron
  • Galvanized finish for enhanced durability
  • Provides a quality solution for general applications
Catalog No. 50-005-30609 Supplier MSC Supplier No. 67328724
This Rigid Eye Single Sheave Pulley is designed for applications utilizing fibrous ropes. It features a 1.5 in outside diameter and is constructed from durable malleable iron with a galvanized finish, ensuring quality performance for various business needs.
  • Designed for use with fibrous ropes
  • Features a 1.5 in outside diameter
  • Constructed from malleable iron
  • Galvanized finish for enhanced durability
  • Provides a quality solution for general applications