Learn More
Promocell GmbH PromoCell Mesenchymal Stem Cell Osteogenic Different. Medium 100 ml, ready-to-use, with C-39813 attached to flask
Catalog No. 50306280
Encompass
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).PromoCell offers five MSC Differentiation Media to efficiently induce differentiation of MSC into adipogenic, chondrogenic (w/ and w/o inducers), osteogenic or neurogenic lineages, respectively.PromoCell Mesenchymal Stem Cell Osteogenic Differentiation Media was developed for the directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into osteogenic lineages.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine cells.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.
Catalog No. 50-306-280 Supplier Promocell GmbH Supplier No. C28013
PromoCell offers a complete Mesenchymal Stem Cell Media System including growth media, differentiation media and human mesenchymal stem cells (MSC).PromoCell offers five MSC Differentiation Media to efficiently induce differentiation of MSC into adipogenic, chondrogenic (w/ and w/o inducers), osteogenic or neurogenic lineages, respectively.PromoCell Mesenchymal Stem Cell Osteogenic Differentiation Media was developed for the directed differentiation of mesenchymal stem cells (MSC) from bone marrow, the umbilical cord matrix (Wharton´s Jelly) and adipose tissue into osteogenic lineages.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine cells.To view a list of references where this medium was used with other species/cell types please check manufacturer homepage.