Learn More
MSC MSC Silicone Sponge Rubber R10470M Roll, 10 yd.
Catalog No. 5000677113
Encompass
This closed cell silicone sponge rubber R10470M roll features an acrylic adhesive backing and is UL approved. It is flexible and suitable for outdoor use, primarily for gasketing applications.
  • Made in USA
  • Acrylic adhesive backing
  • UL approved
  • Closed cell structure
  • Standard grade
  • Flexible
  • Suitable for outdoor use
  • Ideal for gasketing applications
  • Material: silicone sponge rubber R10470M
  • Thickness: 1/8 in.
  • Width: 1 in.
  • Length: 10 yd.
  • Color: orange
  • Shape: rectangle
Catalog No. 50-006-77113 Supplier MSC Supplier No. 20772547
This closed cell silicone sponge rubber R10470M roll features an acrylic adhesive backing and is UL approved. It is flexible and suitable for outdoor use, primarily for gasketing applications.
  • Made in USA
  • Acrylic adhesive backing
  • UL approved
  • Closed cell structure
  • Standard grade
  • Flexible
  • Suitable for outdoor use
  • Ideal for gasketing applications
  • Material: silicone sponge rubber R10470M
  • Thickness: 1/8 in.
  • Width: 1 in.
  • Length: 10 yd.
  • Color: orange
  • Shape: rectangle