Learn More
MSC MSC Stitched Bellows, 24 in, 1-3/4 in ID, 1 ea
Catalog No. 5000803279
Encompass
This stitched bellows is constructed from durable nylon and designed for flexible applications. It features a length of 24 in. and an inside diameter of 1-3/4 in., providing reliable protection in various industrial settings.
  • Made from durable nylon material
  • Features a flexible stitched construction
  • Designed with a 1-3/4 in. inside diameter
  • Measures 24 in. in length
  • Has a material thickness of 0.018 in.
Catalog No. 50-008-03279 Supplier MSC Supplier No. 63414874
This stitched bellows is constructed from durable nylon and designed for flexible applications. It features a length of 24 in. and an inside diameter of 1-3/4 in., providing reliable protection in various industrial settings.
  • Made from durable nylon material
  • Features a flexible stitched construction
  • Designed with a 1-3/4 in. inside diameter
  • Measures 24 in. in length
  • Has a material thickness of 0.018 in.