Learn More
MSC MSC Wire Shelving, Stainless Steel, 25 in Deep, 49 in Wide
Catalog No. 5000534611
Encompass
This wire shelving unit is constructed from stainless steel, offering durability and corrosion resistance. It features dimensions of 25 inches deep and 49 inches wide, providing ample storage space. Designed for use with NuLine units, this shelving is suitable for various industrial and laboratory applications.
  • Constructed from stainless steel for durability and corrosion resistance.
  • Measures 25 inches deep and 49 inches wide for versatile storage.
  • Designed for seamless integration with NuLine units.
Catalog No. 50-005-34611 Supplier MSC Supplier No. 70241468
This wire shelving unit is constructed from stainless steel, offering durability and corrosion resistance. It features dimensions of 25 inches deep and 49 inches wide, providing ample storage space. Designed for use with NuLine units, this shelving is suitable for various industrial and laboratory applications.
  • Constructed from stainless steel for durability and corrosion resistance.
  • Measures 25 inches deep and 49 inches wide for versatile storage.
  • Designed for seamless integration with NuLine units.