Learn More
Medchemexpress LLC HY-135827 5mg Medchemexpress, MSC2360844 CAS:1305267-37-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1358275MG
Encompass_Preferred
Medchemexpress, HY-135827 5mg MSC2360844 CAS:1305267-37-1 MSC2360844 is a potent, orally active and selective PI3Kδ inhibitor, with an IC50 of 145 nM. MSC2360844 shows highly selective against a panel of 278 additional kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1406