Learn More
Selleck Chemical LLC MSC2530818 25mg 1883423-59-3
SDP
Supplier:  Selleck Chemical LLC S838725MG
Encompass_Preferred
MSC2530818, a CDK8 inhibitor with the IC50 of 2.6 nM, displays excellent kinase selectivity, biochemical and cellular potency, microsomal stability, and is orally bioavailable. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1698627