Learn More
TARGETMOL CHEMICALS INC MSC2530818 5MG
SDP
Catalog No. 502578807
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MSC2530818 is an effective selective and orally available CDK8 inhibitor (IC50 2.6 nM). purity: 99%
Catalog No. 50-257-8807 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ02665MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MSC2530818 is an effective selective and orally available CDK8 inhibitor (IC50 2.6 nM). purity: 99%