Learn More
Lonza Walkersville MSCGM Mesenchymal Stem Cell Growth Medium BulletKit
Catalog No. NC9733091
Encompass
Stem cells possess the unique ability to proliferate as an undifferentiated cell type, then, when properly stimulated, differentiate into multiple lineages of cells. Lonza’s MSCGMhuman Mesenchymal Stem Cell Growth BulletKit Medium is a serum containing medium designed to proliferate human bone marrow derived mesenchymal cells in an undifferentiated state.The serum used in Lonza’s MSCGM Human Mesenchymal Stem Cell Growth Medium BulletKit Medium is specifically screened to ensure that the serum neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic, chondrogenic, or adipogenic lineages (when properly stimulated with an appropriate differentiation media).Content:1 x MSCBM Basal Media (PT-3238), 500 mL 1 x MSCGM SingleQuots Growth Supplement Kit (PT-4105)
Catalog No. NC9733091 Supplier Lonza Walkersville Supplier No. PT3001
Stem cells possess the unique ability to proliferate as an undifferentiated cell type, then, when properly stimulated, differentiate into multiple lineages of cells. Lonza’s MSCGMhuman Mesenchymal Stem Cell Growth BulletKit Medium is a serum containing medium designed to proliferate human bone marrow derived mesenchymal cells in an undifferentiated state.The serum used in Lonza’s MSCGM Human Mesenchymal Stem Cell Growth Medium BulletKit Medium is specifically screened to ensure that the serum neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic, chondrogenic, or adipogenic lineages (when properly stimulated with an appropriate differentiation media).Content:1 x MSCBM Basal Media (PT-3238), 500 mL 1 x MSCGM SingleQuots Growth Supplement Kit (PT-4105)