Learn More
Lonza Walkersville MSCGM SingleQuots Supplement
Catalog No. 5000496396
Encompass
The MSCGM hMSC SingleQuots Kit is a component of Lonza s MSCGM BulletKit (catalog no PT-3001) a serum containing medium designed to proliferate human bone marrow derived mesenchymal cells in an undifferentiated state The serum used in Lonza s MSCGM human Mesenchymal Stem Cell Growth Medium BulletKit Medium is specifically screened to ensure that the serum neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic chondrogenic or adipogenic lineages (when properly stimulated with an appropriate differentiation media) Lonza guarantees the performance of Clonetics/Poietics cells only if appropriate Clonetics/Poietics media and reagents are used exclusively and the recommended storage and use protocols are followed Any modifications made to the recommended cell systems including the use of alternative media reagents or protocols will void cell and media performance guarantees
Catalog No. 50-004-96396 Supplier Lonza Walkersville Supplier No. PT4105
The MSCGM hMSC SingleQuots Kit is a component of Lonza s MSCGM BulletKit (catalog no PT-3001) a serum containing medium designed to proliferate human bone marrow derived mesenchymal cells in an undifferentiated state The serum used in Lonza s MSCGM human Mesenchymal Stem Cell Growth Medium BulletKit Medium is specifically screened to ensure that the serum neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic chondrogenic or adipogenic lineages (when properly stimulated with an appropriate differentiation media) Lonza guarantees the performance of Clonetics/Poietics cells only if appropriate Clonetics/Poietics media and reagents are used exclusively and the recommended storage and use protocols are followed Any modifications made to the recommended cell systems including the use of alternative media reagents or protocols will void cell and media performance guarantees