Learn More
New England Biolabs, Inc. MscI – 1250 units
SDP
Catalog No. 50812199
Encompass
A restriction endonuclease that recognizes the sequence TGG_^CCA.
Catalog No. 50-812-199 Supplier New England Biolabs, Inc. Supplier No. R0534L
A restriction endonuclease that recognizes the sequence TGG_^CCA.