Learn More
New England Biolabs, Inc. MscI – 250 units
SDP
Catalog No. 50812201
Encompass
A restriction endonuclease that recognizes the sequence TGG_^CCA.
Catalog No. 50-812-201 Supplier New England Biolabs, Inc. Supplier No. R0534S
A restriction endonuclease that recognizes the sequence TGG_^CCA.