Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ MSCulture Max™ Media

Catalog No. 01001653 Shop All Corning Products
Change view
Click to view available options
Classification:
None
Xeno-free
cGMP, Xeno-free
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Classification
01-001-653 None
01-001-654 Xeno-free
01-001-655 cGMP, Xeno-free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-001-653 Supplier Corning™ Supplier No. 42000KIT
Add to Cart
Add to Cart

High-performance expansion media for batch culture systems

Corning's new MSCulture Max media increase final hMSC yields, streamlines culture, reduces in-process risk, and eliminates media exchanges, enabling higher cell expansion with less media and fewer media changes. The MSCulture Max media consist of a basal medium and supplement with 3 versions available:

Corning MSCulture Max Media, Standard Research-grade

Corning MSCulture Max-XF Media, Xeno-free Research-grade

Corning MSCulture Max-GMP Media, Xeno-free GMP-grade for Bioproduction

Features

  • High-performance expansion media for batch culture systems
  • No media exchanges required
  • No surface coating required

Benefits

  • Unparalleled productivity: Achieves over 10-fold hMSC expansion within 4-5 days without media changes, significantly reducing time and labor costs
  • Transition-friendly formulation:Supports translation from research into clinical manufacturing through research and xeno-free formats. Xeno-free MSCulture Max media is also available in a GMP-grade version.
  • Scalable and efficient bioprocessing: Enables high-volume cell production with minimal media consumption
TRUSTED_SUSTAINABILITY

Specifications

Classification None
Form Liquid
Product Type Cell Culture Media
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives None
Without Additives None
Culture Type Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic), Adherent Cell Culture, Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.