Learn More
Description
Corning's new MSCulture Max media increase final hMSC yields, streamlines culture, reduces in-process risk, and eliminates media exchanges, enabling higher cell expansion with less media and fewer media changes. The MSCulture Max media consist of a basal medium and supplement with 3 versions available:
Corning MSCulture Max Media, Standard Research-grade
Corning MSCulture Max-XF Media, Xeno-free Research-grade
Corning MSCulture Max-GMP Media, Xeno-free GMP-grade for Bioproduction
Features
- High-performance expansion media for batch culture systems
- No media exchanges required
- No surface coating required
Benefits
- Unparalleled productivity: Achieves over 10-fold hMSC expansion within 4-5 days without media changes, significantly reducing time and labor costs
- Transition-friendly formulation:Supports translation from research into clinical manufacturing through research and xeno-free formats. Xeno-free MSCulture Max media is also available in a GMP-grade version.
- Scalable and efficient bioprocessing: Enables high-volume cell production with minimal media consumption
Specifications
Specifications
| Classification | Xeno-free |
| CAS | 67-56-1 |
| Form | Liquid |
| Product Type | Cell Culture Media |
| Serum Level | Serum-free |
| Concentration | 100 μg/mL in methanol (as free base) |
| Sterility | Sterile-filtered |
| Sterilization Method | 0.2 μm Sterile-filtered |
| With Additives | None |
| Without Additives | None |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.