Learn More
System Biosciences LLC MSCV-hspCas9-EF1α-GFP Two Vector Cas9 SmartNuclease™ LentivectorCas9 expression vector for the Two Vector SmartNuclease System (MSCV-hspCas9-EF1α-GFP Lentivector Pre-Packaged Lentiviral Particles = >10^6 IFUs
SDP
Catalog No. NC1899830
Encompass_Preferred
When you want to create stable Cas9 editing cell lines and/or would like to edit the genome of a cell line that is resistant to transfection by plasmids, turn to one of SBI’s Cas9 SmartNuclease Lentivector Systems. Available as lentivector plasmids, ready-to-transduce pre-packaged pseudovirus, and in a package-your-own-lentivector kit with the lentivector plasmid and the LentiStarter 3.0 Packaging Kit, the MSCV-hspCas9-EF1α-GFP Two Vector Cas9 SmartNuclease™ Lentivector expresses human codon-optimized Cas9 wild-type nuclease from the strong MSCV promoter. The vector also includes a GFP marker expressed from the EF1α promoter to verify transduction efficiencies.
Catalog No. NC1899830 Supplier System Biosciences LLC Supplier No. CASLV125VA1
When you want to create stable Cas9 editing cell lines and/or would like to edit the genome of a cell line that is resistant to transfection by plasmids, turn to one of SBI’s Cas9 SmartNuclease Lentivector Systems. Available as lentivector plasmids, ready-to-transduce pre-packaged pseudovirus, and in a package-your-own-lentivector kit with the lentivector plasmid and the LentiStarter 3.0 Packaging Kit, the MSCV-hspCas9-EF1α-GFP Two Vector Cas9 SmartNuclease™ Lentivector expresses human codon-optimized Cas9 wild-type nuclease from the strong MSCV promoter. The vector also includes a GFP marker expressed from the EF1α promoter to verify transduction efficiencies.