Learn More
Meso Scale Discovery MSD GOLD Read Buffer B | 200mL
SDP
Catalog No. NC1948619
Encompass
MSD GOLD Read Buffer B, Read buffer is an essential component of MSD immunoassays: it is required for the generation of electrochemiluminescence (ECL) signals. MSD GOLD Read Buffer B was optimized for use in all MSD assays and can be used as a replacement for MSD GOLD Read Buffer A and MSD Read Buffer T (4x). MSD GOLD Read Buffer B is also recommended for assays that measure whole cell, vesicles, or other membrane-containing analytes. MSD GOLD Read Buffer B has stringent quality control release criteria that ensure lot-to-lot reproducibility. The specifications are designed to meet the requirements of critical assays and longitudinal studies.
Catalog No. NC1948619 Supplier Meso Scale Discovery Supplier No. R60AM3
MSD GOLD Read Buffer B, Read buffer is an essential component of MSD immunoassays: it is required for the generation of electrochemiluminescence (ECL) signals. MSD GOLD Read Buffer B was optimized for use in all MSD assays and can be used as a replacement for MSD GOLD Read Buffer A and MSD Read Buffer T (4x). MSD GOLD Read Buffer B is also recommended for assays that measure whole cell, vesicles, or other membrane-containing analytes. MSD GOLD Read Buffer B has stringent quality control release criteria that ensure lot-to-lot reproducibility. The specifications are designed to meet the requirements of critical assays and longitudinal studies.