Learn More
Medchemexpress LLC HY-108022 10mg Medchemexpress, MSDC-0602 CAS:1133819-87-0 Purity:>98%
SDP
Catalog No. 501968500
Encompass_Preferred
Medchemexpress, HY-108022 10mg MSDC-0602 CAS:1133819-87-0 MSDC-0602, a PPARγ-sparing thiazolidinedione (TZD), interacts with the mitochondrial pyruvate carrier (MPC) and inhibits its activity and are effective for treatment of type 2 diabetes with reducing risk of PPARγ-mediated side effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8500 Supplier Medchemexpress LLC Supplier No. HY10802210MG
Medchemexpress, HY-108022 10mg MSDC-0602 CAS:1133819-87-0 MSDC-0602, a PPARγ-sparing thiazolidinedione (TZD), interacts with the mitochondrial pyruvate carrier (MPC) and inhibits its activity and are effective for treatment of type 2 diabetes with reducing risk of PPARγ-mediated side effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.