Learn More
United States Biological Corporation Mouse Anti-CD209 (CD209 Antigen, C-type Lectin Domain Family 4 Member L, Dendritic Cell-specific ICAM-3-grabbing Non-integrin 1, DC-SIGN, DC-SIGN1, CLEC4L, MGC129965) (MaxLight 490), 100ul
Supplier:  United States Biological Corporation 124578ML490100UL
Encompass_Preferred
MaxLight™490 is a new Blue-Green photostable dye conjugate comparable to DyLight™488, Alexa Fluor™488 and offers better labeling efficiency, brighter imaging and increased immunodetection. Absorbance (491nm); Emission (515nm); Extinction Coefficient 73,000.
Catalog No. NC2867931