Learn More
AbD Serotec Mouse Anti-Human IgG Monoclonal Antibody, Targets CH2 Domain, High Specificity for Human IgG, 0.2 mg
Catalog No. NC1082963
Encompass
Mouse Anti-Human IgG Monoclonal Antibody (Clone R10Z8E9) is designed for precise detection of human IgG in ELISA applications. Its high specificity for the CH2 domain of human IgG, while exhibiting no cross-reactivity with other species, makes it a reliable choice for human-specific immunological studies. With a purified liquid format and a shelf life of 18 months, this antibody ensures reproducible experimental outcomes for therapeutic development.
Catalog No. NC1082963 Supplier AbD Serotec Supplier No. MCA5748G
Mouse Anti-Human IgG Monoclonal Antibody (Clone R10Z8E9) is designed for precise detection of human IgG in ELISA applications. Its high specificity for the CH2 domain of human IgG, while exhibiting no cross-reactivity with other species, makes it a reliable choice for human-specific immunological studies. With a purified liquid format and a shelf life of 18 months, this antibody ensures reproducible experimental outcomes for therapeutic development.