Learn More
Alpha Diagnostic International Mouse Anti-Sm Ig's (total (A+G+M) ELISA Kit, 96 tests, Quantitative
SDP
Supplier:  Alpha Diagnostic International 5405
Encompass
This kit measure anti-Sm Ig's (IgG+A+M) in mouse serum. It is possible to use isotype specific anti-mouse Ig, such as anti-mouse IgM or anti-mouse IgG1-HRP, to measure isotype specific antibodies.
Catalog No. NC1606871