Learn More
Biolegend PE/Dazzle™ 594 Mouse IgG2b, κ isotype Ctrl
Supplier:  Biolegend 400358
Encompass_Preferred
PE/Dazzle™ 594 Mouse IgG2b, κ isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse ; Apps: FC; Size: 100 tests
Catalog No. NC1295072