Learn More
Raybiotech Inc MSE INFLAMMATION ARRAY C1 (4)
Catalog No. 501043235
Encompass_Preferred
RayBio C-Series Mouse Inflammation Antibody Array 1 Kit. Detects 40 Mouse Inflammatory Factors. Suitable for all liquid sample types.
Catalog No. 50-104-3235 Supplier Raybiotech Inc Supplier No. AAMINF14
RayBio C-Series Mouse Inflammation Antibody Array 1 Kit. Detects 40 Mouse Inflammatory Factors. Suitable for all liquid sample types.