Learn More
New England Biolabs, Inc. MseI – 500 units
SDP
Catalog No. 50812181
Encompass
A restriction endonuclease that recognizes the sequence T^TA_A.
Catalog No. 50-812-181 Supplier New England Biolabs, Inc. Supplier No. R0525S
A restriction endonuclease that recognizes the sequence T^TA_A.