Learn More
Medchemexpress LLC α-MSH (11-13) acetate | 2828433-34-5 | >99.3% | 100 MG
SDP
Catalog No. 5000341177
Encompass_Preferred
Alpha-MSH (11-13) acetate functions as a melanocortin-1 receptor ligand and anti-inflammatory agent. It is known to trigger acute intracellular calcium elevation under specific costimulation or pathway inhibition conditions.
  • Inhibits TNF-α-induced NF-κB activation and GPx activity
  • Inhibits HIV replication
  • Inhibits colony formation of *Staphylococcus aureus* and *Candida albicans*
  • Applicable for research into *Staphylococcus aureus* infection, *Candida albicans* infection, and human immunodeficiency virus infection
  • Used in research for traumatic brain injury, corneal epithelial wound, and inflammatory bowel disease
Catalog No. 50-003-41177 Supplier Medchemexpress LLC Supplier No. HY129724A100MG
Alpha-MSH (11-13) acetate functions as a melanocortin-1 receptor ligand and anti-inflammatory agent. It is known to trigger acute intracellular calcium elevation under specific costimulation or pathway inhibition conditions.
  • Inhibits TNF-α-induced NF-κB activation and GPx activity
  • Inhibits HIV replication
  • Inhibits colony formation of *Staphylococcus aureus* and *Candida albicans*
  • Applicable for research into *Staphylococcus aureus* infection, *Candida albicans* infection, and human immunodeficiency virus infection
  • Used in research for traumatic brain injury, corneal epithelial wound, and inflammatory bowel disease