Learn More
Medchemexpress LLC α-MSH (11-13) (acetate) | 2828433-34-5 | 99.3% | 25 MG
SDP
Catalog No. 5000342764
Encompass_Preferred
α-MSH (11-13) acetate is the 11-13 amino acid sequence (lysine-proline-valine) of α-MSH, functioning as a melanocortin-1 receptor ligand and anti-inflammatory agent. It has been shown to reduce fever in rabbits and is intended for research use only.
  • Inhibits TNF-α-induced NF-κB activation and GPx activity
  • Reduces HIV replication and colony formation of S. aureus and C. albicans
  • Applicable to research on infections, traumatic brain injury, and inflammatory bowel disease
  • Facilitates corneal epithelial wound healing
  • Ameliorates DSS-induced colitis and improves recovery in various models
Catalog No. 50-003-42764 Supplier Medchemexpress LLC Supplier No. 25MGHY129724A
α-MSH (11-13) acetate is the 11-13 amino acid sequence (lysine-proline-valine) of α-MSH, functioning as a melanocortin-1 receptor ligand and anti-inflammatory agent. It has been shown to reduce fever in rabbits and is intended for research use only.
  • Inhibits TNF-α-induced NF-κB activation and GPx activity
  • Reduces HIV replication and colony formation of S. aureus and C. albicans
  • Applicable to research on infections, traumatic brain injury, and inflammatory bowel disease
  • Facilitates corneal epithelial wound healing
  • Ameliorates DSS-induced colitis and improves recovery in various models