Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-MSH (11-13) acetate | 2828433-34-5 | 99.3% | 402.49 | C18H34N4O6 | 5 MG
SDP

Catalog No. 5000262205
Encompass_Preferred

α-MSH (11-13) acetate is the acetylated C-terminal tripeptide fragment (Lys-Pro-Val) of alpha-melanocyte stimulating hormone. Supplied as the acetate salt in a white to off-white powder, it is intended for research use and has reported antipyretic and anti-inflammatory activity in animal studies. Handle and store under cold, dry conditions to maintain stability.

  • Contains the C-terminal tripeptide sequence Lys-Pro-Val.
  • Acetate salt supplied as a white to off-white powder.
  • High purity: 99.31% (HPLC).
  • Molecular weight 402.49 and formula C18H34N4O6.
  • Reported antipyretic and anti-inflammatory activity in animal studies.
  • Stable when stored sealed: -80°C up to 2 years, -20°C up to 1 year.

Catalog No. 50-002-62205 Supplier Medchemexpress LLC Supplier No. HY129724A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-MSH (11-13) acetate is the acetylated C-terminal tripeptide fragment (Lys-Pro-Val) of alpha-melanocyte stimulating hormone. Supplied as the acetate salt in a white to off-white powder, it is intended for research use and has reported antipyretic and anti-inflammatory activity in animal studies. Handle and store under cold, dry conditions to maintain stability.

  • Contains the C-terminal tripeptide sequence Lys-Pro-Val.
  • Acetate salt supplied as a white to off-white powder.
  • High purity: 99.31% (HPLC).
  • Molecular weight 402.49 and formula C18H34N4O6.
  • Reported antipyretic and anti-inflammatory activity in animal studies.
  • Stable when stored sealed: -80°C up to 2 years, -20°C up to 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.