Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-MSH (11-13) acetate | 2828433-34-5 | 99.3% | C18H34N4O6 | 10MG
SDP

Catalog No. 5000260150
Encompass_Preferred

α-MSH (11-13) acetate is the acetylated C-terminal tripeptide of α-MSH (sequence Lys-Pro-Val). Supplied for research use, it has reported antipyretic and anti-inflammatory activity in animal studies. The compound is provided as an acetate salt and requires low-temperature, moisture-controlled storage to maintain stability.

  • Sequence: Lys-Pro-Val.
  • CAS number: 2828433-34-5.
  • Molecular weight: 402.49 g·mol⁻¹.
  • Reported purity: 99.3% (reported by distributors).
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: research use only.

Catalog No. 50-002-60150 Supplier Medchemexpress LLC Supplier No. HY129724A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-MSH (11-13) acetate is the acetylated C-terminal tripeptide of α-MSH (sequence Lys-Pro-Val). Supplied for research use, it has reported antipyretic and anti-inflammatory activity in animal studies. The compound is provided as an acetate salt and requires low-temperature, moisture-controlled storage to maintain stability.

  • Sequence: Lys-Pro-Val.
  • CAS number: 2828433-34-5.
  • Molecular weight: 402.49 g·mol⁻¹.
  • Reported purity: 99.3% (reported by distributors).
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended use: research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.