Learn More
CPC Scientific H-Val-Gln-Glu-Ser-Ala-Asp-Gly-Tyr-Arg-Met-Gln-His-Phe-Arg-Trp-Gly-Gln-Pro-Leu-Pro-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MSH004A
Encompass
This peptide containing the melanotropin-core amino acid sequence was isolated from the sea lamprey, Petromuzon marinus. MSH-B was about 10 and 100 times more potent than alpha-MSH and MSH-A, respectively, in a frog skin assay.ONE-LETTER SEQUENCE: VQESADGYRMQHFRWGQPLP-NH2MOLECULAR FORMULA: C107H157N33O29S1MOLECULAR WEIGHT:2401.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [170245-12-2], SYNONYMS: melanotropinsRESEARCH AREA: HormonalREFERENCES: A.Takahashi et al., Int. J. Pept. Protein Res., 46, 197 (1995)
Catalog No. 50-210-9827