Learn More
Cayman Chemical MSH human 10mg
SDP
Catalog No. 502942863
Encompass_Preferred
A peptide hormone; selectively binds to MC1R over MC3R, MC4R, and MC5R (Kis = 0.89, 10.64, 8.18, and 76.9 nM, respectively); induces lipolysis in isolated rabbit adipocytes at 1 µM; increases plasma levels of glucagon, insulin, and free fatty acids in fed or fasted rabbits at 45 UG/animal; CSF levels are increased in patients with restless legs syndrome with painful symptoms
Catalog No. 50-294-2863 Supplier Cayman Chemical Supplier No. 4154910 MG
A peptide hormone; selectively binds to MC1R over MC3R, MC4R, and MC5R (Kis = 0.89, 10.64, 8.18, and 76.9 nM, respectively); induces lipolysis in isolated rabbit adipocytes at 1 µM; increases plasma levels of glucagon, insulin, and free fatty acids in fed or fasted rabbits at 45 UG/animal; CSF levels are increased in patients with restless legs syndrome with painful symptoms