Learn More
Med Vet International Optilene Surgical Mesh 15cm X 15cm, Sterile, J1247D, Each
Catalog No. 5000495762
Encompass_Preferred
• Monofilament Polypropylene nonabsorbable • Larger pore size • Blue guidelines in material helps proper placement • More elastic mesh • Can be used on minimally invasive surgeries Specifications: Weight 60g/m2 Thickness 0.53mm Pore Size 1.5mm Optilene Mesh 15.0cm x 15.0cm
Catalog No. 50-004-95762 Supplier Med Vet International Supplier No. JOR1247D
• Monofilament Polypropylene nonabsorbable • Larger pore size • Blue guidelines in material helps proper placement • More elastic mesh • Can be used on minimally invasive surgeries Specifications: Weight 60g/m2 Thickness 0.53mm Pore Size 1.5mm Optilene Mesh 15.0cm x 15.0cm