Learn More
Med Vet International Optilene Surgical Mesh 20cm x 30cm, Sterile, J1247B, Each
Catalog No. 5000495659
Encompass_Preferred
• Monofilament Polypropylene nonabsorbable • Larger pore size • Blue guidelines in material helps proper placement • More elastic mesh • Can be used on minimally invasive surgeries Specifications: Weight 60g/m2 Thickness 0.53mm Pore Size 1.5mm Optilene Mesh 20.0cm x 30.0cm
Catalog No. 50-004-95659 Supplier Med Vet International Supplier No. JOR1247B
• Monofilament Polypropylene nonabsorbable • Larger pore size • Blue guidelines in material helps proper placement • More elastic mesh • Can be used on minimally invasive surgeries Specifications: Weight 60g/m2 Thickness 0.53mm Pore Size 1.5mm Optilene Mesh 20.0cm x 30.0cm