Learn More
Medchemexpress LLC α-MSH TFA | 171869-93-5 | 98.54% | 5 MG
SDP
Catalog No. 5000425749
Encompass_Preferred
α-MSH TFA (α-Melanocyte-Stimulating Hormone) is an endogenous neuropeptide and a melanocortin receptor 4 (MC4R) agonist. It exhibits anti-inflammatory and antipyretic activities and is a post-translational derivative of pro-opiomelanocortin (POMC).
  • Endogenous neuropeptide
  • Acts as a melanocortin receptor 4 (MC4R) agonist
  • Exhibits anti-inflammatory activities
  • Exhibits antipyretic activities
  • Modulates central nervous system inflammation by acting directly on melanocortin receptors in glial cells
  • Modulates NFκB activation and inhibits translocation of transcription factor κB to the nucleus
  • Effective in modulating inflammatory reactions in vivo
Catalog No. 50-004-25749 Supplier Medchemexpress LLC Supplier No. HYP0252A5MG
α-MSH TFA (α-Melanocyte-Stimulating Hormone) is an endogenous neuropeptide and a melanocortin receptor 4 (MC4R) agonist. It exhibits anti-inflammatory and antipyretic activities and is a post-translational derivative of pro-opiomelanocortin (POMC).
  • Endogenous neuropeptide
  • Acts as a melanocortin receptor 4 (MC4R) agonist
  • Exhibits anti-inflammatory activities
  • Exhibits antipyretic activities
  • Modulates central nervous system inflammation by acting directly on melanocortin receptors in glial cells
  • Modulates NFκB activation and inhibits translocation of transcription factor κB to the nucleus
  • Effective in modulating inflammatory reactions in vivo