Learn More
Electron Microscopy Sciences MeshWell Plate 12-well with Cluster
SDP
Catalog No. 5030870
Encompass_Preferred
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening. Includes one MeshWell and one Cluster plate.
Catalog No. 50-308-70 Supplier Electron Microscopy Sciences Supplier No. 6470212
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500um (0.0197") opening. Includes one MeshWell and one Cluster plate.